3-phenyl-1H-indole-2-carboxylic acid 97%

Warning

Synonyms: 3-Phenylindole-2-carboxylic acid

H33246-MD H33246-03 H33246-MD H33246-03
AAH33246-MDEA 80.78 USD
AAH33246-MD CAAAH33246-MD CAAAH33246-03 AAH33246-03
3-phenyl-1H-indole-2-carboxylic acid 97%
3-phenyl-1H-indole-2-carboxylic acid
Formula: C₁₅H₁₁NO₂
MW: 237.258 g/mol
Melting Pt: 195…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00974212
CAS Number: 6915-67-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR