Diethyl-10-undecyn-1-ylphosphonate 95%
|
Warning
|
Synonyms:
10-Undecyn-1-ylphosphonic acid diethyl ester
H33242-MD
H33242-MD
H33242-03
H33242-03
AAH33242-MDEA
168.72
USD
CAAAH33242-MD
CAAAH33242-03
AAH33242-MD
AAH33242-03
Diethyl-10-undecyn-1-ylphosphonate 95%
Diethyl-10-undecyn-1-ylphosphonate
Formula:
C₁₅H₂₉O₃P MW: 288.37 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17012685 CAS Number: 1242248-76-5 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...