4-Chloro-3-ethoxy-2-fluorophenylacetonitrile 97%

Warning

Synonyms: 4-Chloro-3-ethoxy-2-fluorophenylacetonitrile

H33224-03 H33224-03 H33224-MD H33224-MD
AAH33224-MDEA 89.81 USD
AAH33224-MD CAAAH33224-03 CAAAH33224-MD AAH33224-03
4-Chloro-3-ethoxy-2-fluorophenylacetonitrile 97%
4-Chloro-3-ethoxy-2-fluorophenylacetonitrile
Formula: C₁₀H₉ClFNO
MW: 213.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687187
CAS Number: 1323966-15-9
UN: 3439
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR