3,5-Dichloro-1H-indazole 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33220-03
H33220-03
H33220-MD
H33220-MD
AAH33220-03EA
357.99
USD
AAH33220-03
CAAAH33220-MD
CAAAH33220-03
AAH33220-MD
3,5-Dichloro-1H-indazole 95%
3,5-Dichloro-1H-indazole
Formula:
C₇H₄Cl₂N₂ Melting Pt: 240…242 °C Storage Temperature: Ambient |
MDL Number:
MFCD09263200 CAS Number: 36760-20-0 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...