Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-3-methyl-5-(trifluoromethyl)aniline ≥97%

Supplier: Alfa Aesar
Danger
Synonyms: 4-Fluoro-3-methyl-5-(trifluoromethyl)aniline

H33212-06 H33212-06 H33212-03 H33212-03
AAH33212-06EA 515 USD InStock
CAAAH33212-06 AAH33212-06 CAAAH33212-03 AAH33212-03
4-Fluoro-3-methyl-5-(trifluoromethyl)aniline ≥97%
4-Fluoro-3-methyl-5-(trifluoromethyl)aniline


Formula: C₈H₇F₄N
MW: 193.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201067
CAS Number: 1373920-72-9
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty