Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-methyl-5-(trifluoromethyl)benzoyl chloride ≥97%

Supplier: Alfa Aesar
Danger

H33182-03 H33182-06 H33182-03 H33182-06
AAH33182-06EA 269.86 USD InStock
CAAAH33182-03 AAH33182-06 AAH33182-03 CAAAH33182-06
3-methyl-5-(trifluoromethyl)benzoyl chloride ≥97%
3-methyl-5-(trifluoromethyl)benzoyl chloride


Formula: C₉H₆ClF₃O
MW: 222.591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD1631600
CAS Number: 261952-09-4
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty