4'-Methoxybiphenyl-4-carboxaldehyde 96%

Warning

Synonyms: 4'-Methoxy-[1,1'-biphenyl]-4-carbaldehyde

H33166-03 H33166-06 H33166-03 H33166-06
AAH33166-06EA 237.54 USD
AAH33166-06 AAH33166-03 CAAAH33166-06 CAAAH33166-03
4'-Methoxybiphenyl-4-carboxaldehyde 96%
4'-Methoxybiphenyl-4-carboxaldehyde
Formula: C₁₄H₁₂O₂
MW: 212.248 g/mol
Melting Pt: 98…100 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01862517
CAS Number: 52988-34-8

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR