3-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid 95%

Warning

Synonyms: 2-Carboxy-3-chloroimidazo[1,2-a]pyridine

H33162-MD H33162-03 H33162-03 H33162-MD
AAH33162-MDEA 72.3 USD
CAAAH33162-MD AAH33162-MD AAH33162-03 CAAAH33162-03
3-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid 95%
3-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Formula: C₈H₅ClN₂O₂
MW: 196.591 g/mol
Melting Pt: 203…205 °C
Storage Temperature: Ambient
MDL Number: MFCD09836169
CAS Number: 100001-79-4

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR