3-Methoxy-2-(trifluoromethyl)benzyl bromide 97%

Danger

H33160-03 H33160-MD H33160-03 H33160-MD
AAH33160-03EA 198.69 USD
AAH33160-03 CAAAH33160-MD AAH33160-MD CAAAH33160-03
3-Methoxy-2-(trifluoromethyl)benzyl bromide 97%
3-Methoxy-2-(trifluoromethyl)benzyl bromide
Formula: C₉H₈BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD18398595
CAS Number: 1261828-76-5
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR