2-Fluoro-5-methoxy-3-(trifluoromethyl)benzamide 97%

Warning

Synonyms: 2-Fluoro-5-methoxy-3-(trifluoromethyl)benzamide

H33148-03 H33148-06 H33148-03 H33148-06
AAH33148-06EA 410.7 USD
CAAAH33148-06 AAH33148-06 AAH33148-03 CAAAH33148-03
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzamide 97%
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzamide
Formula: C₉H₇F₄NO₂
MW: 237.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201042
CAS Number: 1373920-76-3

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR