4-bromo-1-methyl-1H-pyrazole-3-carbonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-1-methyl-1H-pyrazole-3-carbonitrile

H33131-03 H33131-MD H33131-MD H33131-03
AAH33131-03EA 294.99 USD
CAAAH33131-03 CAAAH33131-MD AAH33131-03 AAH33131-MD
4-bromo-1-methyl-1H-pyrazole-3-carbonitrile 97%
4-bromo-1-methyl-1H-pyrazole-3-carbonitrile
Formula: C₅H₄BrN₃
MW: 186.012 g/mol
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00103545
CAS Number: 287922-71-8

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR