7-Bromobenzo[b]furan 97%

Warning

Synonyms: 7-Bromobenzofuran

H33129-MD H33129-03 H33129-03 H33129-MD
AAH33129-03EA 186.47 USD
CAAAH33129-MD AAH33129-03 CAAAH33129-03 AAH33129-MD
7-Bromobenzo[b]furan 97%
7-Bromobenzo[b]furan
Formula: C₈H₅BrO
MW: 197.031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09056756
CAS Number: 133720-60-2

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR