Imidazo[1,2-a]pyridine-6-carboxylic acid 95%

Warning

H33128-03 H33128-06 H33128-03 H33128-06
AAH33128-03EA 127.42 USD
AAH33128-03 CAAAH33128-06 AAH33128-06 CAAAH33128-03
Imidazo[1,2-a]pyridine-6-carboxylic acid 95%
Imidazo[1,2-a]pyridine-6-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 244…247 °C
Storage Temperature: Ambient
MDL Number: MFCD07021498
CAS Number: 139022-25-6

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR