4-Methoxy-3-(trifluoromethoxy)phenylacetonitrile 97%

Warning

Synonyms: 4-Methoxy-3-(trifluoromethoxy)phenylacetonitrile

H33126-03 H33126-MD H33126-MD H33126-03
AAH33126-MDEA 88.24 USD
AAH33126-MD CAAAH33126-MD AAH33126-03 CAAAH33126-03
4-Methoxy-3-(trifluoromethoxy)phenylacetonitrile 97%
4-Methoxy-3-(trifluoromethoxy)phenylacetonitrile
Formula: C₁₀H₈F₃NO₂
MW: 231.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18398529
CAS Number: 853771-93-4
UN: 3276
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR