3-Methoxy-4-(trifluoromethyl)benzamide 97%

Warning

H33121-06 H33121-06 H33121-03 H33121-03
AAH33121-03EA 123.21 USD
CAAAH33121-03 CAAAH33121-06 AAH33121-03 AAH33121-06
3-Methoxy-4-(trifluoromethyl)benzamide 97%
3-Methoxy-4-(trifluoromethyl)benzamide
Formula: C₉H₈F₃NO₂
Melting Pt: 219…220 °C
Storage Temperature: Ambient
MDL Number: MFCD13185679
CAS Number: 1214328-36-5

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR