5-Chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde 97%

Warning

H33065-MD H33065-03 H33065-MD H33065-03
AAH33065-03EA 167.61 USD
AAH33065-03 AAH33065-MD CAAAH33065-03 CAAAH33065-MD
5-Chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde 97%
5-Chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde
Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD22682805
CAS Number: 1823049-90-6

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR