4'-(Pentafluorothio)acetophenone 97%

Warning

H33056-06 H33056-03 H33056-06 H33056-03
AAH33056-06EA 277.5 USD
AAH33056-06 AAH33056-03 CAAAH33056-03 CAAAH33056-06
4'-(Pentafluorothio)acetophenone 97%
4'-(Pentafluorothio)acetophenone
Formula: C₈H₇F₅OS
MW: 246.19
Storage Temperature: Ambient
MDL Number: MFCD16652408
CAS Number: 401892-83-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR