3,5-Difluoro-2-methylbenzamide 97%
|
Warning
|
H33022-MD
H33022-03
H33022-03
H33022-MD
AAH33022-03EA
236.43
USD
CAAAH33022-MD
AAH33022-03
CAAAH33022-03
AAH33022-MD
3,5-Difluoro-2-methylbenzamide 97%
3,5-Difluoro-2-methylbenzamide
Formula:
C₈H₇F₂NO Storage Temperature: Ambient |
MDL Number:
MFCD19687171 CAS Number: 1323966-37-5 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...