3-Methoxy-2-(trifluoromethyl)benzaldehyde 97%
|
Warning
|
H33018-03
H33018-MD
H33018-MD
AAH33018-MDEA
41.22
USD
AAH33018-MD
CAAAH33018-03
CAAAH33018-MD
3-Methoxy-2-(trifluoromethyl)benzaldehyde 97%
3-Methoxy-2-(trifluoromethyl)benzaldehyde
Formula:
C₉H₇F₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD13185677 CAS Number: 1214384-89-0 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...