2,3-Dichloro-6-fluoroanisole 97%
|
Warning
|
H32016-03
H32016-06
H32016-03
H32016-06
AAH32016-06EA
228.66
USD
CAAAH32016-06
CAAAH32016-03
AAH32016-06
AAH32016-03
2,3-Dichloro-6-fluoroanisole 97%
2,3-Dichloro-6-fluoroanisole
Formula:
C₇H₅Cl₂FO MW: 195.02 g/mol Melting Pt: 47…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD09832260 CAS Number: 1017777-48-8 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...