2-Methoxythiazole 98%

Warning

B25655-06 B25655-14 B25655-06 B25655-14
AAB25655-14EA 263.07 USD
AAB25655-14 AAB25655-06 CAAAB25655-06 CAAAB25655-14
2-Methoxythiazole 98%
2-Methoxythiazole
Formula: C₄H₅NOS
MW: 115.16 g/mol
Boiling Pt: 150…151 °C
Flash Pt: 49 °C(120 °F)
Storage Temperature: Ambient
MDL Number: MFCD01631143
CAS Number: 14542-13-3
UN: 1993
ADR: "3",III

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR