Sodium antimonyl-L-tartrate ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12707-18
A12707-0B
A12707-18
A12707-30
A12707-0B
A12707-30
AAA12707-0BEA
384.99
USD
AAA12707-0B
CAAAA12707-0B
CAAAA12707-30
AAA12707-18
CAAAA12707-18
AAA12707-30
Sodium antimonyl-L-tartrate ≥98%
Sodium antimonyl-L-tartrate
Formula:
C₈H₄Na₂O₁₂Sb₂ MW: 581.60 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00065402 CAS Number: 34521-09-0 EINECS: 252-070-9 UN: 1549 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...