Would you like to log in again?
Amersham CyDye Monoclonal Antibody Labeling Kits, GE Healthcare
Supplier: GE Healthcare - Life Sciences
CY™
PA32001 PA35001 PA33001 PA35001 PA33001 PA32001
95017-386EA 459.96 USD InStock
95017-386 95017-394 95017-390 CA95017-394L CA95017-390L CA95017-386L
Amersham CyDye Monoclonal Antibody Labeling Kits, GE Healthcare
Protein Linking and Labeling Systems
Conjugations are carried out under mild conditions and are complete within 1 h.
  • Convenient way to tag antibodies with CyDye Fluorescent Dyes
Dyes are packaged in premeasured amounts and foil-sealed to ensure consistent labelings.
CyDye fluorescent dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.
Monoclonal antibody kits contain all reagents required for 2 × 0.1 mg reactions. No wetted part.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty