Would you like to log in again?
Amersham ECL Direct Labeling Module, GE Healthcare
Supplier: GE Healthcare - Life Sciences
ECL™
RPN3005 RPN3005
95038-274EA 585.54 USD InStock
CA95038-274L 95038-274
Amersham ECL Direct Labeling Module, GE Healthcare
Transfer Membranes
ECL Direct Nucleic Acid Labeling and Detection Systems are based on the direct labeling of DNA or RNA probes with horseradish peroxidase (HRP) in a simple 20 minute chemical reaction
  • Sufficient for labeling 5 µg nucleic acid and detecting 2000 cm² of membrane
  • Optimal storage conditions are 2 to 8 °C
The resulting probe can be used without purification. Detection is achieved by generation of light via the HRP-catalyzed breakdown of luminol.
Delivery information: Each system contains labeling reagent, crosslinker, control DNA, blocking agent, ECL Detection Reagents, and ECL Gold Hybridization Buffer.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty