Would you like to log in again?
CDP-Star Detection Reagents, GE Healthcare
Supplier: GE Healthcare - Life Sciences
JACKSON SAFETY®
RPN3682 RPN3683 RPN3683 RPN3682
95038-294EA 494.4 USD InStock
CA95038-292L CA95038-294L 95038-294 95038-292
CDP-Star Detection Reagents, GE Healthcare
Transfer Membranes
AlkPhos Direct Labeling and Detection Systems are based on the rapid, direct labeling of DNA or RNA probes with thermostable alkaline phosphatase
DNA or RNA probes are labeled during a 30 minute reaction and resulting probes can be used without purification. Hybridization stringency can be controlled by temperature as well as salt concentration. Detection is rapid, taking only 30 minutes from completion of hybridization to application of detection reagent.
Each system includes the following reagents, sufficient for detection of 2500 cm² membrane: labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer, blocking agent, CDP-Star or ECF detection reagents.
CDP-Star or ECF detection can be used.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty