Would you like to log in again?
Adaptor Kit, GE Healthcare
Supplier: GE Healthcare - Life Sciences
80-6105-27 80-6105-27
10035-994EA 38.01 USD InStock
CA10035-994 10035-994
Adaptor Kit, GE Healthcare
Fittings
Connects 4 mm output jack to 4 mm high voltage leads.
Note that this adaptor kit is not CE compliant.
Ordering information: Supplied with Allen wrench.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty