Would you like to log in again?
Tubing Connector, 1/8", GE Healthcare
Supplier: GE Healthcare - Life Sciences
18-1121-17 18-1121-17
10498-006EA 166.96 USD InStock
CA10498-006 10498-006
Tubing Connector, 1/8", GE Healthcare
Connectors Connectors, Tubing
Tubing connector is constructed from polyetheretherketone (PEEK)
Connector features a 5/16" male connection for 1/8" O.D. tubing. Black, for use with tubing used on AKTA systems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty