Would you like to log in again?
XK Empty Chromatography Columns, GE Healthcare
Supplier: GE Healthcare - Life Sciences
19011401 19011301 18100084 19011501 18100085 19031501 19014501
101724-942EA 337.62 USD InStock
101724-942 101724-940 101724-826 101724-944 101724-828 101724-954 101724-946
XK Empty Chromatography Columns, GE Healthcare
Chromatography Columns
XK Column Tubes can be used in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules with the following few exceptions - chlorinated hydrocarbons, acetone and other ketones, aliphatic esters and phenol
Solutions containing more than 10% NaOH, 10% HCl, other strong mineral acids or 5% acetic acid should not be used.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty