Would you like to log in again?
Superdex Columns, Peptide Precision, GE Healthcare
Supplier: GE Healthcare - Life Sciences
Superdex™
29-0362-31
10048-752EA 2461.49 USD InStock
10048-752
Superdex Columns, Peptide Precision, GE Healthcare
Chromatography Columns
Superdex™ Peptide PC 3.2/30 is a prepacked Precision Column for high-resolution gel filtration of peptides and other small biomolecules with molecular weights between 100 and 7000, in the micropreparative scale.
  • Excellent choice for the polishing step in a purification procedure
  • Precision columns (PC) for micropurification and analysis: Sample volumes of 4 - 50 µl
  • Easy and predictable scale-up
  • Excellent reproducibility and durability
  • Hardware designed for convenient direct connection to high performance chromatography systems
  • Flow rate of 0,01 ml/min to 0,15 ml/min
Caution: Note: Not suitable for use with ÄKTAprime plus system.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty