Would you like to log in again?
Whatman PZ 001 Tweezers, GE Healthcare
Supplier: GE Healthcare - Whatman
10477602 10477602
11008-548EA 47.94 USD InStock
CA11008-548 11008-548
Whatman PZ 001 Tweezers, GE Healthcare
Forceps
The stainless steel forceps with smooth angled jaws (104 mm long) are highly suitable for handling membrane filters
They are autoclavable and can be flame-sterilized with ethanol.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty