Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

illustra plasmidPrep Midi Flow Kit, GE Healthcare

Supplier: GE Healthcare - Life Sciences
28-9042-68
102853-560EA 2605.92 USD InStock
102853-560
illustra plasmidPrep Midi Flow Kit, GE Healthcare
Nucleic Acid Reagents Sanger Sequencing Reagents
illustra plasmidPrep Midi Flow Kit can be used to process bacterial sample volumes of 25 to 50 ml for high copy number plasmid DNA and up to 500 ml for low copy number plasmid DNA

  • Provides plasmid DNA with excellent purity and low endotoxin levels
  • Sample type E. coli cultures containing high, low or very low-copy number plasmids and cosmid vectors up to 50 kbp
  • Modified alkaline lysis procedure
  • Anion exchange chromatography
  • > 70% (strain-dependent) of supercoiled pDNA
  • \< 10 endotoxin units (EU) per μg of plasmid DNA
  • Increased ease of use due to minimal pipetting volume changes, color-coded kit components, and pre-equilibrated columns
  • Purification format is single-use, gravity flow illustra Fast-Flow plasmid 250 columns
  • Prep time 215 minutes

High column capacity allows for a proportional yield for input sample volume of 25 to 50 ml (for high copy number plasmid) and up to 500 ml for low copy number plasmid. Used For the rapid extraction and purification of high-quality plasmid DNA from medium scale cultures for use in transfection, sequencing, and enzymatic amplification and modification

Kit consistently produces high yields of pure plasmid DNA with low levels of endotoxin contamination.

The kit consists of a high-performance, anion exchange purification medium that combines excellent flow characteristics with an exceptional capacity and specificity for plasmid DNA. The kit contains fast-flow plasmid 250 columns, resuspension, lysis, neutralization, wash, elution, and TE buffers, and a full protocol booklet with a detachable, quick reference protocol card.

Ordering information: Kit contains lysis buffer type 11 (Red, 2 x 360 mL), lysis buffer type 12 (Clear, 2 x 360 mL), lysis buffer type 13 (Blue, 2 x 360 mL), wash buffer type 2 (Yellow, 5 x 480 mL), elution buffer type 3 (Silver, 480 mL), TE buffer type 1 (Green, 120 mL), illustra Fast-Flow plasmid 250 columns (2 x 50), column stand, waste reservoir and adapters (12).

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty