HR (High Resolution) Columns, Cytiva
Supplier: Cytiva
18146001
101724-890EA 1375.95 USD
101724-890
HR (High Resolution) Columns, Cytiva
HR 16 columns are designed for high resolution liquid chromatography of biomolecules performed at low to medium pressures. The columns are suitable for use with high performance media such as Sepharose High Performance and SOURCE medium. Column hardware consists of borosilicate glass, ETFE, PP, PEEK, EPDM and PTFE.
  • Compatible with any high performance system including ÄKTA design systems
  • Provide precision sample application and elution and excellent physical and chemical stability
  • Glass construction, with no metal parts, allows visual observation at all times
  • Resistant to most solutions used in liquid chromatography except hydrocarbons, aromatic solvents and chlorinated hydrocarbons
  • Bed support: PP
Wide range of accessories, adapters and replacement parts available.