Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman Grade GF 8 Glass Microfiber Filters with Binder, GE Healthcare

Supplier: GE Healthcare - Whatman

Whatman™
10370172 10370119 10370105 10370105 10370119 10370172
10034-836PK 117.88 USD InStock
CA11013-858 10034-836 CA11013-420 10034-834 CA11013-562 10034-838
Whatman Grade GF 8 Glass Microfiber Filters with Binder, GE Healthcare
Filters Glass Fiber Filters
GE’s Whatman Grade GF 8 glass microfiber filters are used for filtration of coarse particles. GF 8 offers faster filtration and a higher air flow rate compared to the similar Grade GF 9 filters.

  • Nominal particle retention of 3 µm
  • Air flow rate of 12 s/100 ml/1.56 cm²
  • Inorganic binder for increased strength and temperature resistant up to 500° C
  • Fast filtration speed
  • Can be used as a pre-filter for dense samples
  • Function as air pollution filters for detecting furans, PCB, and other chemicals

Environmental pollution analyses often require binder-strengthened glass filters to support heavy samples

With the added strength of an inorganic binder, grade GF 8 glass microfiber filters serve as air pollution filters by retaining a range of heavy particles found in vapor samples. Other applications include: Air pollution filters for Polychlorinated Biphenyl (PCB), Dichlorodiphenyldichloroethylene(DDE), Dichlorodiphenyltrichloroethane (DDT), furans, and dioxin retention for analysis; Pollution measurements in industrial, urban, and populated areas; Workplace dust measurements; Measurement of dust fraction in technical gases; Dust-collecting effectiveness testing.

Several options are available in the Whatman range of glass filters with binder from GE Healthcare’s Life Sciences business.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty