Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

C Columns, GE Healthcare

Supplier: GE Healthcare - Life Sciences
19510301 19500201 19510101 19520301 19500301 19520101 19510201 19520201 19510401 19500101
101724-996EA 388.02 USD InStock
101724-996 101724-978 101724-992 101725-110 101724-980 101725-106 101724-994 101725-108 101724-998 101724-976
C Columns, GE Healthcare
Chromatography Columns
C-series columns are designed for standard chromatography. The column hardware consists of borosilicate glass, PA, fuororubber, PP, superpolyoxymethylene and silicon rubber. The eluent inlet pieces incorporate a “reverse funnel” design that distributes eluent evenly over the cross-sectional area of the gel bed surface. This causes minimal disturbance of the gel bed and results in improved flow properties. These columns are resistant to aqueous solutions and solvents used in liquid chromatography. They are not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, NaOH, >10% HCl, >5% acetic acid, or strong mineral acid.

  • Economical modular columns for low pressure or gravity feed, providing even distribution of eluent to the medium bed, even without flow adapters
  • Compatible with a variety of bulk media
  • Flow adapters are available for each of the column diameters to allow optimal bed heights for each application, as are thermostatic jackets for precise temperature control
  • Packing reservoirs for each column diameter optimize packing conditions
  • Porosity: 10 or 80 µm
  • Bed support: PA net
  • Inlet and outlet ID: 1,0 mm

Flow adapters: The basic C columns are of fixed volume, but can be upgraded by adding flow adaptors (which keep bed height constant and protect from insoluble particles) and a thermostatic jacket (see below). Three adjustable flow adaptors are available for the C column series (AC 10, AC 16 and AC 26), allowing the choice of bed height to suit the application.

Gel and eluent reservoirs: RC packing reservoirs, which are available for all three C column types, are quick and easy to assemble. Curved walls minimise the separation of gel beads by size during packing to produce homogeneous columns. Once the gel is in the column, the RC reservoir can be connected to a pump or Mariotte flask to give optimal flow rates for gel packing.

Temperature control: When working with temperature dependent separations, resolution and recovery can often be increased by fitting a C column with a JC thermostatic jacket. This allows accurate control or separation temperature without a cold room. JC jackets increase mechanical resistance of C columns, enabling operating pressures of upto 1 bar.

Delivery information: Supplied with PE tubing (1,0 mm ID, 2 m in length), bed support screens, snap-on net rings and instructions.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty