Would you like to log in again?
Whatman Grade 17 Chr Cellulose Chromatography Papers, GE Healthcare
Supplier: GE Healthcare - Whatman
3017-820 3017-820 3017-915 3017-621 3017-917 3017-8793 3017-621 3017-917 3017-8793
10035-440PK 104.06 USD InStock
CA11013-604 10035-440 21427-393 10035-438 CA28297-452 10035-842 CA11013-770 28297-452 CA11013-766
Whatman Grade 17 Chr Cellulose Chromatography Papers, GE Healthcare
Filters Filter Paper Chromatography Paper
Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
  • Also widely used in protein and nucleic acid blotting
Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
A thick (0.92 mm) and highly absorbent paper with a very high flow rate of 190 mm/30 min. Suitable for the heaviest loadings and an excellent choice for preparative paper chromatography and electrophoresis.
Note: This data shown may be based on historic Whatman records. As is normal practice, customers should determine and validate for themselves (as necessary) that the products are suitable for their specific application
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty