Would you like to log in again?
Whatman HEPA-CAP™ 150 Venting Filters, GE Healthcare
Supplier: GE Healthcare - Whatman
6702-9500 6702-9500
14230-540EA 104.62 USD InStock
14230-540 CA14230-540
Whatman HEPA-CAP™ 150 Venting Filters, GE Healthcare
Filters Venting Filters
HEPA-CAP high-efficiency particulate air filters from GE are robust, efficient, glass fiber filters designed to support high particle retention, high flow rate applications.
  • Glass filter media is strengthened by dual lamination with a tough polyester monofilament
  • Typical retention efficiency of 99.97% of all particles ≥ 0.3 µm in air
  • Depth filter design allows for high loading capacity and bidirectional flow
  • Appropriate for tissue culture applications
GE Healthcare’s Life Sciences business offers a variety of venting filters
HEPA-CAP air filters are used throughout the scientific, research, and industrial environments in a variety of air and gas filtration applications requiring high particle retention and the ability to capture dirt and debris at high flow rates.
Whatman HEPA-CAP filters exhibit high-efficiency particulate retention, retaining 99.97% of all particles ≥ 0.3 µm in air.
In addition to HEPA air filters, PolyVENT and SteriVENT options are available.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty