Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nafion® 5% (w/w) in water/1-propanol ≥1.00 meq/g ion exchange capacity

Supplier: BeanTown Chemical


225940-500ML 225940-25ML 225940-100ML
BT225940-500MLEA 619.44 USD InStock
BT225940-500ML BT225940-25ML BT225940-100ML
Nafion® 5% (w/w) in water/1-propanol ≥1.00 meq/g ion exchange capacity
Nafion®
CAS: 31175-20-9; MDL No: MFCD00132790
UN No: UN1987; Haz Class: 3; Packing Group: III
Liquid
Flash point: 24°C (75°F)
Density (g/mL): 0.93


Formula: C₉HF₁₇O₅S
MW: 544,135 g/mol
MDL Number: MFCD00132790
CAS Number: 31175-20-9
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty