Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium hydride ≥98% (trace metals basis excluding Mg)

Supplier: BeanTown Chemical

Synonyms: Calcium dihydride

225920-250G 225920-50G
BT225920-250GEA 568.63 USD InStock
BT225920-250G BT225920-50G
Calcium hydride ≥98% (trace metals basis excluding Mg)
Calcium hydride
CAS: 7789-78-8; EC No: 232-189-2; MDL No: MFCD00010897
UN No: UN1404; Haz Class: 4.3; Packing Group: I
-10 Mesh Powder; Molecular Formula: CaH2; MW: 42.09
Melting Point: 190°
Moisture Sensitive


Formula: H₂Ca
MW: 42,09 g/mol
Melting Pt: 816 °C
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00010897
CAS Number: 7789-78-8
EINECS: 232-189-2
UN: 1404
ADR: 4.3,I
Merck Index: 13,01674
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty