Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium metavanadate ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Lithium vanadium oxideLithium vanadium trioxideLithium vanadate

225865-250G 225865-50G
BT225865-250GEA 259.05 USD InStock
BT225865-250G BT225865-50G
Lithium metavanadate ≥99.9% (trace metals basis)
Lithium metavanadate
CAS: 15060-59-0; EC No: 239-124-7; MDL No: MFCD00053997
UN No: UN3285; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: LiVO3; MW: 105.88


Formula: LiO₃V
MW: 105,88 g/mol
Melting Pt: 605 °C
Density: 2.74 g/cm³ (20 °C)
MDL Number: MFCD00053997
CAS Number: 15060-59-0
EINECS: 239-124-7
UN: 3285
ADR: 6.1,III
Order Now
Specification Test Results
Assay 99.9%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty