Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile ≥99.7% for HPLC

Supplier: BeanTown Chemical

Danger
Synonyms: Methane, cyano-Ethanoic acid nitrileMethyl cyanideMethanecarbonitrileEthanenitrileCyanomethaneACN

225835-4L 225835-1L 225835-4X4L 225835-4X1L
BT225835-4LEA 220.78 USD InStock
BT225835-4L BT225835-1L BT225835-4X4L BT225835-4X1L
Acetonitrile ≥99.7% for HPLC
Acetonitrile
CAS: 75-05-8; EC No: 200-835-2; MDL No: MFCD00001878; RTECS: AL7700000
UN No: UN1648; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3CN; MW: 41.05
Melting Point: -48°; Boiling Point: 81-82°; Flash point: 2°C (36°F)
Density (g/mL): 0.786; Refractive Index: 1.344


Formula: H₃CCN
MW: 41,05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://us.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
Assay 99.7% min.


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty