Hexachloro-1,3-butadiene ≥97%

225770-250G 225770-50G 225770-1KG
BT225770-50GEA 43.2 USD
BT225770-50G BT225770-250G BT225770-1KG
Hexachloro-1,3-butadiene ≥97%
Hexachloro-1,3-butadiene

  • RTECS: EJ0700000
  • Refractive index: 1.555
  • Liquid

Formula: C₄Cl₆
MW: 260.76 g/mol
Boiling Pt: 210…220 °C
Melting Pt: –21 °C
Density: 1.665 g/mL
MDL Number: MFCD00000836
CAS Number: 87-68-3
EINECS: 201-765-5
UN: 2279
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR