Magnesium ≥99%, turnings

Supplier: BeanTown Chemical

225730-100G 225730-500G 225730-2.5KG
BT225730-2.5KGEA 321.2 USD
BT225730-2.5KG BT225730-500G BT225730-100G
Magnesium ≥99%, turnings
Magnesium

  • Solid

Formula: Mg
MW: 24.3 g/mol
Boiling Pt: 1090 °C
Melting Pt: 648 °C
Density: 1.74 g/mL
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR