Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-2,4-dinitrobenzene ≥98%

Supplier: BeanTown Chemical


225715-25G 225715-100G
BT225715-25GEA 66.88 USD InStock
BT225715-25G BT225715-100G
1-Bromo-2,4-dinitrobenzene ≥98%
1-Bromo-2,4-dinitrobenzene
CAS: 584-48-5; EC No: 209-539-8; MDL No: MFCD00041873; RTECS: CY9021500
UN No: UN3459; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C6H3BrN2O4; MW: 247.00
Melting Point: 71-73°


Formula: C₆H₃BrN₂O₄
MW: 247 g/mol
Melting Pt: 75 °C
Density: Min. 1 g/cm³ (20 °C)
MDL Number: MFCD00041873
CAS Number: 584-48-5
EINECS: 209-539-8
UN: 3459
ADR: 6.1,III
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty