Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Hydroxy-3-methylbutanone ≥90%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: 3-Hydroxy-3-methyl-2-butanone3-Hydroxy-3-methylbutan-2-one

225675-5G 225675-100G 225675-25G
BT225675-5GEA 32.11 USD InStock
BT225675-5G BT225675-100G BT225675-25G
3-Hydroxy-3-methylbutanone ≥90%, Technical Grade
3-Hydroxy-3-methylbutanone
CAS: 115-22-0; EC No: 204-073-1; MDL No: MFCD00004460
UN No: UN1224; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C5H10O2; MW: 102.13
Boiling Point: 140-141°; Flash point: 42°C (108°F)
Density (g/mL): 0.971; Refractive Index: 1.415


Formula: C₅H₁₀O₂
MW: 102,13 g/mol
Boiling Pt: 140 °C (1013 hPa)
Density: 0.9595 g/cm³ (13 °C)
Storage Temperature: ambient
MDL Number: MFCD00004460
CAS Number: 115-22-0
EINECS: 204-073-1
UN: 1224
ADR: 3,III
Order Now
Specification Test Results
Assay 90%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty