Chloropentacarbonylrhenium ≥98%

Supplier: BeanTown Chemical

Synonyms: Pentacarbonylchlororhenium(I)Chloropentacarbonylrhenium(I)Rhenium pentacarbonyl chloride

225655-1G
BT225655-1GEA 181.77 USD
BT225655-1G
Chloropentacarbonylrhenium ≥98%
Chloropentacarbonylrhenium

  • Air Sensitive, Moisture Sensitive
  • Crystalline

Formula: C₅ClO₅Re
MW: 361.71 g/mol
MDL Number: MFCD00013296
CAS Number: 14099-01-5
EINECS: 237-948-1
UN: 3466
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR