2-Chloro-2-methylpropane ≥98%

Synonyms: tert-Butyl chloride

225625-250ML 225625-2.5L 225625-500ML
BT225625-250MLEA 30 USD
BT225625-250ML BT225625-500ML BT225625-2.5L
2-Chloro-2-methylpropane ≥98%
2-Chloro-2-methylpropane

  • RTECS: TX5040000
  • Refractive index: 1.385
  • Hygroscopic
  • Liquid

Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 50…52 °C
Melting Pt: –25 °C
Density: 0.851 g/mL
Flash Pt: –23 °C (–9 °F)
MDL Number: MFCD00000816
CAS Number: 507-20-0
EINECS: 208-066-4
UN: 1127
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR