3-Furaldehyde ≥97% stabilized

Synonyms: Furan-3-carbaldehyde

225555-5G 225555-100G 225555-25G
BT225555-5GEA 66.12 USD
BT225555-5G BT225555-100G BT225555-25G
3-Furaldehyde ≥97% stabilized
3-Furaldehyde

  • Refractive index: 1.493
  • Air Sensitive, Light Sensitive
  • Liquid

Formula: C₅H₄O₂
MW: 96.09 g/mol
Boiling Pt: 144 °C/732 mmHg
Density: 1.111 g/mL
Flash Pt: 48 °C (118 °F)
MDL Number: MFCD00010424
CAS Number: 498-60-2
ADR: 6.1 (3),II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR