Phenyl acrylate ≥97%

Supplier: BeanTown Chemical

225490-25G 225490-5G
BT225490-5GEA 72.41 USD
BT225490-5G BT225490-25G
Phenyl acrylate ≥97%
Phenyl acrylate

  • Refractive index: 1.521
  • Liquid

Formula: C₉H₈O₂
Boiling Pt: 89…90 °C/12 mmHg
Density: 1.076 g/mL
MDL Number: MFCD00048145
CAS Number: 937-41-7
EINECS: 213-329-1
ADR: 9,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR