Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isoprene ≥99% stabilized

Synonyms: 2-Methyl-1,3-butadiene2-Methylbuta-1,3-diene

225445-100ML 225445-500ML
BT225445-100MLEA 26.2 USD InStock
BT225445-100ML BT225445-500ML
Isoprene ≥99% stabilized
Isoprene
CAS: 78-79-5; EC No: 201-143-3; MDL No: MFCD00008600; RTECS: NT4037000
UN No: UN1218; Haz Class: 3; Packing Group: I
Liquid; Molecular Formula: C5H8; MW: 68.12
Melting Point: -146°; Boiling Point: 34°; Flash point: -54°C (-65°F)
Density (g/mL): 0.681; Refractive Index: 1.422


Formula: C₅H₈
MW: 68,12 g/mol
Boiling Pt: 34 °C (1013 hPa)
Melting Pt: -146 °C
Density: 0.681 g/cm³ (20 °C)
Flash Pt: -48 °C
MDL Number: MFCD00008600
CAS Number: 78-79-5
EINECS: 201-143-3
UN: 1218
ADR: 3,I
Merck Index: 13,05220
Order Now
Specification Test Results
Assay 99%

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty