4,4,5,5-Tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: BeanTown Chemical

Synonyms: Pinacol Borane

225340-25G 225340-5G
BT225340-25GEA 188.48 USD
BT225340-25G BT225340-5G
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane ≥97%
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane

  • Refractive index: 1.396
  • Air Sensitive
  • Liquid

Formula: C₆H₁₃BO₂
MW: 127.98 g/mol
Boiling Pt: 42…43 °C/50 mmHg
Density: 0.882 g/mL
Flash Pt: 5 °C (41 °F)
MDL Number: MFCD00674030
CAS Number: 25015-63-8
ADR: 4.3 (3),II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR